Learn More
Medchemexpress LLC Sonlicromanol | 1541170-75-5 | 99.8% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY12157750MG
Encompass_Preferred
Sonlicromanol (KH176) is an orally active reactive oxygen species (ROS) modulator used in preclinical and clinical research to study mitochondrial disease. It is supplied for research use only and is typically provided as a small research pack for laboratory studies.
  • Orally active small-molecule ROS modulator
  • Modulates reductive and oxidative cellular distress with anti-inflammatory effects
  • Used to investigate mitochondrial disease mechanisms and redox biology
  • Shown to protect mitochondrial-diseased cells from redox stress in published studies
  • Provided as a research reagent, not for therapeutic use
  • Molecular formula C19H28N2O3, molecular weight 332.4 g/mol
Catalog No. 50-002-34781