Learn More
Medchemexpress LLC Sonolisib | 502632-66-8 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN67755MG
Encompass_Preferred
Sonolisib (PX-866), an improved Wortmannin analogue, is an oral, irreversible, and pan-isoform inhibitor of PI3K. It inhibits the production of the secondary messenger phosphatidylinositol-3,4,5-trisphosphate (PIP3) and activation of the PI3K/Akt signaling pathway, which may result in inhibition of tumor cell growth and survival in susceptible tumor cell populations. It exhibits antitumor activity.
  • Oral, irreversible, and pan-isoform inhibitor of PI3K
  • Inhibits production of secondary messenger phosphatidylinositol-3,4,5-trisphosphate (PIP3)
  • Inhibits activation of PI3K/Akt signaling pathway
  • Exhibits antitumor activity
  • May inhibit tumor cell growth and survival
Catalog No. 50-004-21138