Learn More
Medchemexpress LLC Sonrotoclax | 2383086-06-2 | 99.61% | 890.10 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY148026
Encompass_Preferred
Sonrotoclax is a potent, orally active Bcl2 inhibitor that exhibits an effective cell-killing effect against various lymphoma and leukemia cell lines. It demonstrates antineoplastic activity and inhibits tumor growth in relevant models.
  • Potent, orally active Bcl2 inhibitor
  • Effective cell-killing effect against various lymphoma and leukemia cell lines
  • Exhibits antineoplastic activity
  • Inhibits tumor growth
  • For research use only
Catalog No. 50-003-62631