Learn More
Enterprise Technology Solutions Sontara EC Engineered Cloths
SDP
Supplier:  Enterprise Technology Solutions HOSPR811
Encompass_Preferred
For wet or dry use.More versatile than cloth rags and more durable than paper.Low cost per use.Solvent resistant and exceptionally low lint.
Catalog No. 50-211-1166