Learn More
ABS Imaging Systems Sony High Glossy Printing Paper, case of 10
SDP
Supplier:  ABS Imaging Systems UPP110HG10PK
Encompass_Preferred
High density, high glossy thermal print paper. Prints are about 25% glossier than the UPP-110HD media. Approximately 270 prints per roll, 10 rolls per case.
Catalog No. NC1649809