Learn More
Medchemexpress LLC HY-N2284 1mg Medchemexpress, Sophoflavescenol CAS:216450-65-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN22841MG
Encompass_Preferred
Medchemexpress, HY-N2284 1mg Sophoflavescenol CAS:216450-65-6 Sophoflavescenol is a prenylated flavonol, which shows great inhibitory activity with IC 50 of 0.013 μM against Phosphodiesterase 5 (PDE5) , and also inhibits RLAR, HRAR, AGE, BACE1, AChE and BChE with IC 50 s of 0.30 µM, 0.17 µM, 17.89 µg/mL, 10.98 µM, 8.37 µM and 8.21 µM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3901