Learn More
TARGETMOL CHEMICALS INC SOPHORAFLAVANONE G 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T5S100010MG
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Sophoraflavanone G (Kushenol F) (Kushenol F) isolated from Sophora flavescens induces MDA-MB-231 and HL-60 cells apoptosis through suppression of MAPK-related pathways. purity: 99%
Catalog No. 50-257-9715