Learn More
Cayman Chemical Sophoraflavnon G 10mg
SDP
Supplier:  Cayman Chemical 3475510 MG
Encompass_Preferred
A flavonoid; active against 21 strains of MRSA (MICs = 3.13-6.25 UG/ml) and inhibits the activities of α-glucosidase and β-amylase (Kis = 5.6 and 30.6 µM, respectively); cytotoxic to HL-60 and HepG2 cancer cells (IC50s = 12.5 and 13.3 µM, respectively) and induces apoptosis in HL-60 cells at 10 or 25 µM; reduces LPS-induced PGE2 production in RAW 264.7 cells from 1-50 µM; inhibits carrageenan-induced paw edema in rats
Catalog No. 50-291-1375