Learn More
Medchemexpress LLC Sophoranone | 23057-55-8 | 5 MG

Supplier: Medchemexpress LLC HYN25295MG
Sophoranone is a flavonoid compound that can be isolated from the root of *Sophora subprostrata*. It exhibits anti-inflammatory activity in mouse RAW264.7 cells by inhibiting LPS-induced NO production.
- Flavonoid compound
- Isolated from the root of Sophora subprostrata
- Exhibits anti-inflammatory activity
- Molecular weight: 460.60
- Formula: C30H36O4
- Appearance: White to off-white solid
- Purity: 99.67%
- Soluble in DMSO (100 mg/mL)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.