Learn More
Medchemexpress LLC HY-N0423 100mg Medchemexpress, Sophoricoside CAS:152-95-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN0423100MG
Encompass_Preferred
Medchemexpress, HY-N0423 100mg Sophoricoside CAS:152-95-4 Sophoricoside is an isoflavone glycoside isolated from Sophora japonica and has anti-inflammatory, anti-cancer and immunosuppressive effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2789