Learn More
Selleck Chemical LLC Sophoricoside S3776-25mg
SDP
Supplier:  Selleck Chemical LLC S377625MG
Encompass_Preferred
Sophoricoside (SOPH) is an isoflavone isolated from Sophora japonica (Leguminosae) with numerous pharmacological effects including anti- inflammatory anti-cancer and immunosuppressive effects
Catalog No. 50-310-9625