Learn More
Medchemexpress LLC Sorafenib | 284461-73-0 | MFCD06411450 | 99.7% | 464.83 | C21H16ClF3N4O3 | 50 MG

Supplier: Medchemexpress LLC HY10201R50MG
Sorafenib (Standard) is an analytical reference standard of the multikinase inhibitor sorafenib intended for research and analytical applications. It is provided as a 50 mg packaged standard for use in method validation, assay calibration, and quantitative analysis.
- Purity 99.71% for reliable reference measurements.
- Suitable for method development and assay calibration.
- Chemical formula: C21H16ClF3N4O3; molecular weight: 464.83.
- CAS number 284461-73-0 for unambiguous identification.
- Intended for laboratory and analytical use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.