Learn More
Selleck Chemical LLC Sorafenib tosylate S1040-10mM/1mL
SDP
Supplier:  Selleck Chemical LLC S104010MM/1ML
Encompass_Preferred
Sorafenib tosylate is a multikinase inhibitor of Raf-1 and B-Raf with IC50 of 6 nM and 22 nM in cell-free assays respectively Sorafenib Tosylate inhibits VEGFR-2 VEGFR-3 PDGFR- Flt-3 and c-KIT with IC50 of 90 nM 20 nM 57 nM 59 nM and 68 nM respectively Sorafenib Tosylate induces autophagy and apoptosis and activates ferroptosis with anti-tumor activity
Catalog No. 50-310-8464