Learn More
Medchemexpress LLC Sorbic acid | 110-44-1 | 112.13 | 500 G

Supplier: Medchemexpress LLC HYN0626500G
Sorbic acid is an effective, safe, and non-toxic food preservative that can inhibit most molds, yeasts, and certain bacteria. It is orally active and can enhance the growth performance of economically important animals by activating the insulin-like growth factor system (IGF). It can also lead to lipid accumulation by disrupting lipid clearance pathways.
- Effective, safe, and non-toxic food preservative.
- Inhibits most molds, yeasts, and certain bacteria.
- Orally active.
- Can enhance growth performance in certain animals by activating the IGF system.
- Can lead to lipid accumulation by disrupting lipid clearance pathways.
- Purity: 99.99%.
- Appearance: Solid.
- Color: White to off-white.
- Structure classification: Ketones, aldehydes, acids.
- Initial source: Plants other families, microorganisms, endogenous metabolite.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.