Learn More
Medchemexpress LLC Sorbifolin | 23130-22-5 | 300.26 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN115525MG
Encompass_Preferred
Sorbifolin, a flavone glucoside, can be isolated from the Pterogyne nitens. Sorbifolin has myeloperoxidase inhibitory and radical scavenging activities. Sorbifolin is also a MPO inhibitor with an IC50 value of 19.2 nM.
  • Flavone glucoside
  • Can be isolated from Pterogyne nitens
  • Myeloperoxidase inhibitory activity
  • Radical scavenging activities
  • MPO inhibitor with an IC50 value of 19.2 nM
Catalog No. 50-004-15724