Learn More
Medchemexpress LLC HY-50289 1mg Medchemexpress, Sorbinil CAS:68367-52-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY502891MG
Encompass_Preferred
Medchemexpress, HY-50289 1mg Sorbinil CAS:68367-52-2 Sorbinil, is an Aldose reductase inhibitor ( ARI ).Sorbinil plays therapeutic role in treating diabetes and diabetic complications, decreases AR activity and inhibits polyol pathway, it to be found comparatively safer than other ARIs for human use [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2868