Learn More
Cayman Chemical SordarInsodIum salt 5mg
SDP
Supplier:  Cayman Chemical 262555 MG
Encompass_Preferred
An inhibitor of fungal protein synthesis; binds to EF-2 in the presence of ribosomes; inhibits the uncoupled GTPase activity of equimolar mixtures of EF-2 and ribosomes from C. albicans (IC50 = 0.1 μM); inhibits protein synthesis in cell-free lysates of C. albicans, C. glabrata, and C. neoformans (IC50s = 0.01, 0.2, and 0.06 μg/ml, respectively) but not in rabbit reticulocytes (IC50 = >100 μg/ml); inhibits the growth of C. albicans (MIC = 8 μg/ml) but not C. glabrata or C. neoformans (MICs = >125 μg/ml)
Catalog No. 50-293-6974