Learn More
Sigma Aldrich Fine Chemicals Biosciences SorensonTM low binding aer
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z7194394800EA
Encompass
Tips feature a new surface technology designed for quantitative assays in which sample binding can significantly alter results or for critical small volume protocols that require a high degree of accuracy. Ideal for molecular biology applications such as PCR. Inert hydrophobic barrier prevents pipettor contamination.
Catalog No. 50-290-9002