Learn More
Medchemexpress LLC Sorivudine | 77181-69-2 | 99.7% | 349.13 | C11H13BrN2O6 | 1MG
SDP
Supplier:  Medchemexpress LLC HY1230321MG
Encompass_Preferred
Sorivudine is a synthetic pyrimidine nucleoside antiviral reagent (BV-araU) used in biochemical and virology research as a thymidine analogue that interferes with viral DNA synthesis. The compound is supplied with batch documentation and analytical reports for research use; consult the product documentation for handling and storage instructions.
  • Synthetic pyrimidine nucleoside antiviral reagent.
  • Thymidine analogue that inhibits viral DNA synthesis.
  • Molecular formula C11H13BrN2O6; molecular weight 349.13.
  • Reported purity 99.69%.
  • Available in small research sizes (1 mg, 5 mg, 10 mg, 50 mg).
  • Analytical documentation available: data sheet, SDS, COA.
Catalog No. 50-002-35732