Learn More
Medchemexpress LLC Sortilin/SORT1 Huma 5ug | 5ug
SDP
Supplier:  Medchemexpress LLC HYP787735UG
Encompass_Preferred
The Sortilin/SORT1 protein acts as a sorting receptor in the Golgi apparatus and as a clearance receptor on the cell surface It plays a crucial role in protein transport to lysosomes independent of M6PR Sortilin/SORT1 Protein Human (HEK293 His) is the recombinant human-derived Sortilin/SORT1 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-94843