Learn More
Medchemexpress LLC Sortin1 | 503837-98-7 | 99.5% | 10 MG
SDP
Supplier:  Medchemexpress LLC HY1282710MG
Encompass_Preferred
Sortin1 is a vacuolar protein sorting inhibitor that targets carboxypeptidase Y (CPY). It is a soluble and membrane vacuolar marker molecule found in plants and yeast.
  • Targets carboxypeptidase Y (CPY).
  • Functions as a vacuolar protein sorting inhibitor.
  • Soluble and membrane vacuolar marker molecule.
  • Suitable for research in plants and yeast.
Catalog No. 50-003-39671