Learn More
Selleck Chemical LLC Sotagliflozin (LX4211) 25mg 1018899-04-1
SDP
Supplier:  Selleck Chemical LLC S810325MG
Encompass_Preferred
Sotagliflozin (LX4211) is an oral dual SGLT1/SGLT2 inhibitor with IC50 of 36 nM and 1.8 nM, respectively. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5656