Learn More
Apexbio Technology LLC Sotalol 3930-20-9 50mg
SDP
Supplier:  Apexbio Technology LLC B136750
Encompass_Preferred
Sotalol (CAS 3930-20-9) is a non-selective adrenergic beta-receptor antagonist that inhibits both 1- and 2-adrenergic signaling pathways By blocking these receptors sotalol reduces sympathetic stimulation of the myocardium leading to decreased heart rate and myocardial contractility Furthermore it exhibits class III antiarrhythmic properties by prolonging cardiac action potential duration through inhibition of potassium channels Sotalol is utilized in biomedical research for investigating mechanisms underlying arrhythmogenesis and for evaluating therapeutic strategies in models of life-threatening ventricular and supraventricular arrhythmias
Catalog No. 50-000-88700