Learn More
Medchemexpress LLC Sotalol-d6 hydrochloride | 1246820-85-8 | 99.0% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0437AS1MG
Encompass_Preferred
Sotalol-d6 hydrochloride is a deuterium labeled Sotalol hydrochloride. It acts as an orally active, non-selective competitive β-adrenergic receptor blocker and a potent antiarrhythmic agent, primarily used in the research of pediatric arrhythmias. The compound functions by blocking β-receptors and potassium KCNH2 channels.
  • Deuterium labeled compound
  • Non-selective competitive β-adrenergic receptor blocker
  • Potent antiarrhythmic agent
  • Blocks β-receptors and potassium KCNH2 channels
  • Useful as a tracer in quantitative analysis
  • Serves as an internal standard for NMR, GC-MS, or LC-MS analysis
  • Solid appearance
  • White to off-white color
Catalog No. 50-004-09168