Learn More
Medchemexpress LLC Sotalol (hydrochloride) | 959-24-0 | 99.9% | 1 ML
Supplier: Medchemexpress LLC HYB043710MM/1ML
Sotalol hydrochloride is an orally active, non-selective β-adrenergic receptor blocker and a potent antiarrhythmic agent. It is used for the research of pediatric arrhythmias. Sotalol hydrochloride blocks β-receptors and potassium KCNH2 channels, and acts as an Antiepileptic Agent.
- Orally active
- Non-selective β-adrenergic receptor blocker
- Potent antiarrhythmic agent
- Blocks β-receptors
- Blocks potassium KCNH2 channels
- Acts as an antiepileptic agent
- Used for pediatric arrhythmias research