Learn More
Medchemexpress LLC Soticlestat | 1429505-03-2 | 99.83% | 373.45 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY109123100MG
Encompass_Preferred
Soticlestat (TAK-935; OV935) is a first-in-class, potent, selective, and orally active cholesterol 24-hydroxylase (CH24H) inhibitor with potential for epilepsy syndromes research.
  • It is for research use only, not intended for human use.
  • It inhibits the catalytic activity of human cholesterol 24-hydroxylase (CH24H) in a concentration-dependent manner with an IC50 of 4.5 nM.
  • Treatment with soticlestat lowers brain 24S-hydroxycholesterol in a dose-dependent manner.
  • It substantially reduces premature deaths of APP/PS1-Tg mice at a dose lowering brain 24S-hydroxycholesterol by approximately 50%.
  • It can suppress potassium-evoked extracellular glutamate elevations in the hippocampus.
Catalog No. 50-003-44346