Learn More
Medchemexpress LLC Sotorasib racemate | 5MG
SDP
Supplier:  Medchemexpress LLC HY114277A5MG
Encompass_Preferred
Sotorasib racemate (Compound A) is an orally active racemate of Sotorasib (HY-114277) a covalent inhibitor of KRAS G12C mutant which induces adaptive feedback activation of MAPK pathway Sotorasib racemate also exerts inhibitor activity against KRAS G12C induced cancer and can be applied to cancer research[1]
Catalog No. 50-002-27009