Learn More
Medchemexpress LLC HY-12768 10mg , Sotuletinib CAS:953769-46-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1276810MG
Encompass_Preferred
HY-12768 10mg Sotuletinib CAS: 953769-46-5 Sotuletinib (BLZ945) is a potent, selective and brain-penetrant CSF-1R (c-Fms) inhibitor with an IC50 of 1 nM, showing more than 1,000-fold selectivity against its closest receptor tyrosine kinase homologs. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0013