Learn More
Medchemexpress LLC HY-132840 5mg , Sovilnesib CAS:2410796-79-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1328405MG
Encompass_Preferred
HY-132840 5mg Sovilnesib CAS: 2410796-79-9 Sovilnesib (AMG 650) is a kinesin-like protein KIF18A inhibitor (WO2020132648). Sovilnesib can be used for the research of cancer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9491