Learn More
Selleck Chemical LLC Sovleplenib E4580-5MG
SDP
Supplier:  Selleck Chemical LLC E45805MG
Encompass_Preferred
Sovleplenib(HMPL-523) is a highly potent and selective SYK inhibitor with IC50 of 0 025 M that exhibits anti-tumor activities in vitro and in vivo Sovleplenib exhibits synergistic activities in combination with BTK PI3K and Bcl2 family inhibitors in killing human-diffused large B cell lymphoma (DLBCL)
Catalog No. 50-000-39069