Learn More
Genetex Inc SOX9 Monoclonal Antibody [MSVA-709R] HistoMAX™ (GTX04459) GeneTex - IHC-Optimized Antibody.
SDP
Supplier:  Genetex Inc GTX04459
Encompass_Preferred
The SOX9 antibody [MSVA-709R] HistoMAX™ (GTX04459) is a Rabbit Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human mucosa from duodenum tissue using GTX04459 SOX9 antibody [MSVA-709R] HistoMAX. Strong nuclear SOX9 staining of crypt epithelial cells. The SOX9 staining intensity markedly decreases towards the surface epithelium and the tips of the villosities remain SOX9 negative.
Catalog No. NC3667741