Learn More
Avanti Polar Lipids L-α-Phosphatidylinositol (Soy) (sodium salt), powder, 50 mg, purity >99% (TLC), light sensitive, hygroscopic, stable for 3 months at -20°C
Supplier: Avanti Polar Lipids 840044P50MG
L-α-Phosphatidylinositol (Soy) (sodium salt), known as Soy PI, is a high-purity phosphoinositide crucial for various biochemical and cellular research applications. This phospholipid, derived from soy, is supplied as a hygroscopic, light-sensitive powder with a purity exceeding 99%. It plays a significant role in studying membrane dynamics, signal transduction pathways, and cholesterol metabolism, making it essential for liposome preparation. When stored properly at -20°C, it remains stable for three months.
- High purity over 99% (TLC)
- Hygroscopic nature
- Light sensitive
- Insoluble in ethanol and DMSO
- Soluble in Chloroform:Methanol:Water (65:25:4) at 5 mg/mL
- Packing size of 50 mg
- Stored at -20°C for optimal stability
- Shipping condition: Dry ice
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.