Learn More
Medchemexpress LLC Soya lecithin | 8030-76-0 | 5 G
Supplier: Medchemexpress LLC HY1546445G
Soya Lecithin is a phospholipid mixture that can be used as a drug delivery vehicle and is a pharmaceutical excipient. It can form a lipid bilayer structure through self-assembly, and its binding properties are amphiphilic (hydrophilic head and hydrophobic tail), encapsulating hydrophobic drugs.
- Forms stable nanoliposomes or microemulsions
- Improves solubility and cellular uptake efficiency of poorly soluble drugs
- Enhances drug delivery
- Regulates cell proliferation
- Promotes proliferation of rat bone marrow mesenchymal stem cells