Learn More
Medchemexpress LLC Soya Lecithin | 8030-76-0 | 50 G
SDP
Supplier:  Medchemexpress LLC HY15464450G
Encompass_Preferred
Soya Lecithin is a phospholipid mixture that functions as a drug delivery vehicle and a pharmaceutical excipient. It can self-assemble to form a lipid bilayer structure with amphiphilic properties, possessing both a hydrophilic head and a hydrophobic tail. This characteristic allows it to encapsulate hydrophobic drugs and form stable nanoliposomes or microemulsions, improving the solubility and cellular uptake efficiency of poorly soluble drugs.
  • Functions as a drug delivery vehicle and pharmaceutical excipient
  • Forms lipid bilayer structures through self-assembly
  • Amphiphilic properties, enabling encapsulation of hydrophobic drugs
  • Forms stable nanoliposomes or microemulsions
  • Improves solubility and cellular uptake efficiency of poorly soluble drugs
  • Enhances drug delivery and regulates cell proliferation
  • Promotes proliferation of rat bone marrow mesenchymal stem cells (rBMSCs)
Catalog No. 50-003-76816