Learn More
Medchemexpress LLC Soyasaponin ab | 118194-13-1 | 99.2% | 1437.52 | C67H104O33 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN302610MG
Encompass_Preferred
Soyasaponin Ab is a soy-derived triterpenoid saponin provided for research use as a biochemical modulator of PPARγ activity. It is reported to be orally active, can inhibit PPARγ transcriptional activity, and may induce apoptosis at higher concentrations. Consult the SDS and batch COA for handling, solubility, and lot-specific purity information.
  • Orally active soy-derived triterpenoid saponin.
  • Inhibits PPARγ transcriptional activity.
  • Used in cell-based assays probing PPAR signaling and metabolic pathways.
  • Available in small research pack sizes (e.g., 5 mg-100 mg).
  • Typical HPLC purity ~99.2%; confirm batch COA for lot-specific purity.
  • Solid powder, soluble in DMSO for assay preparation.
  • Supplied with SDS and COA for safety and analytical data.
Catalog No. 50-002-89389