Learn More
Selleck Chemical LLC SP-13786 5mg
SDP
Supplier:  Selleck Chemical LLC S08425MG
Encompass_Preferred
SP-13786 (UAMC-1110)(compound 60) is a potent and selective inhibitor of fibroblast activation protein (FAP) and prolyl oligopeptidase (PREP) with IC50 of 3.2 nM and 1.8 ?M, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3050