Learn More
Abcam Sp-5,6-Dichloro-cBIMPS, cAMP-dependent protein kinase activator, 1MG
Supplier:  Abcam AB1452021MG
Encompass_Preferred
MW 419.2 g/mol, Purity >98%. cAMP analog. Cell-permeable, potent activator of cAMP-dependent protein kinase (PKA; Ka = 30 nM); Selective for PKA over cGMP-dependent protein kinase (PKG; Ka = 10 μM). Not significantly hydrolyzed by type I, II or III phosphodiesterases.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5400