Learn More
OTT Hydromet Kipp & Zonen - SP LITE 2 PYRANOMETER 5M CABLE
Supplier:  OTT Hydromet 0339920001
Encompass_Preferred
SP Lite2 is designed for routine measurement of solar radiation. It is especially designed for Photovoltaic / solar energy module monitoring. SP Lite2 can be used under all weather conditions. The sensor measures the solar energy received from the entire hemisphere. It is ideal for measuring available energy for use in solar energy applications, plant growth, thermal convection and evapotranspiration. SP Lite2 uses a photodiode detector, which creates a voltage output that is proportional to the incoming radiation. Also due to the unique design of the diffuser, its sensitivity is proportional to the cosine of the angle of incidence of the incoming radiation, allowing for accurate and consistent measurements. SP Lite2 is easy to use. It can be directly connected to voltmeter or data logger. Direct readout in Watts per square meter (W/m²) can be derived from the measured voltage divided by the calibration coefficient. It is fitted with the Kipp & Zonen high quality yellow cable.
Catalog No. NC1919641