Learn More
TARGETMOL CHEMICALS INC SP2509 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T230425MG
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SP2509 is a specific histone demethylase LSD1 inhibitor(IC50 =13 nM). purity: 99%
Catalog No. 50-258-0899