Learn More
Medchemexpress LLC Sp4206 | 515846-21-6 | 97.0% | 1 MG

Supplier: Medchemexpress LLC HY1194241MG
SP4206 is a small-molecule inhibitor of the interleukin-2 (IL-2) / IL-2Rα interaction; it binds IL-2 with high affinity (Kd = 70 nM) and blocks IL-2 binding to IL-2Rα (Kd = 10 nM). It is supplied as a solid for in vitro research applications.
- Binds IL-2 with high affinity (Kd = 70 nM).
- Blocks IL-2 binding to IL-2Rα (Kd = 10 nM).
- Molecular formula C30H37Cl2N7O6 and molecular weight 662.56 g·mol⁻¹.
- Supplied as 1 mg solid suitable for research use.
- Reported purity approximately 97.0% (HPLC).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.