Learn More
Medchemexpress LLC Sp4206 | 515846-21-6 | 97.0% | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1194245MG
Encompass_Preferred
Small-molecule research inhibitor of the IL-2/IL-2Rα protein-protein interaction. It binds IL-2 with high affinity (Kd = 70 nM) and prevents IL-2 binding to IL-2Rα (Kd = 10 nM). Provided as a high-purity solid or DMSO solution for in vitro and in vivo studies; molecular formula C30H37Cl2N7O6; molecular weight 662.56; purity ~97%.
  • Inhibits IL-2/IL-2Rα interaction; binds IL-2 with Kd 70 nM.
  • Blocks IL-2 binding to IL-2Rα (reported Kd 10 nM).
  • High purity (~96.97%) suitable for research applications.
  • Available as solid quantities and 10 mM DMSO solution for ready-to-use formulations.
  • Soluble in DMSO (100 mg/mL); multiple in vivo formulation protocols reported.
Catalog No. 50-002-32149