Learn More
Medchemexpress LLC HY-12041 500mg , SP600125 CAS:129-56-6 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY12041500MG
Encompass_Preferred
Medchemexpress, HY-12041 500mg SP600125 CAS:129-56-6 Formula:C14H8N2O IC50: 40/40/90 nM (JNK1/2/3) Purity:98% SP600125 is a broad-spectrum JNK inhibitor for JNK1 , JNK2 and JNK3 with IC 50 of 40 nM, 40 nM and 90 nM, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2328