Learn More
Cayman Chemical SP600125ngatIve control 1mg
SDP
Supplier:  Cayman Chemical 188581 MG
Encompass_Preferred
A methylated analog of SP600125 that has much lower affinity for JNK isoforms (IC50s = 18 and 24 µM for JNK2 and JNK3, respectively)
Catalog No. 50-298-1858