Learn More
ELMI NORTH AMERICA SPACER FOR 3-10ML VACUTNR(1EA)
Supplier:  ELMI NORTH AMERICA CM6MT7
Encompass_Preferred
Spacer for 3-10ml vacutainers . Compatible with Rotors 6M.05, 6M.02, 8M.05 and 8M.02 (1 Each)
Catalog No. NC2124870