Learn More
Medchemexpress LLC HY-P1422A 1mg Medchemexpress, Spadin (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP1422A1MG
Encompass_Preferred
Medchemexpress, HY-P1422A 1mg Spadin (TFA) CAS: Spadin TFA, a natural peptide derived from a propeptide released in blood, is able to block the TREK-1 (KCNK2 or K2P2.1) channel activity. Spadin TFA binds specifically to TREK-1 with an affinity of 10 nM. Spadin TFA is an efficient antidepressant in mice. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3054