Learn More
Andwin Scientific SPAN-8 P1000 STANDARD NS 480CS
SDP
Supplier:  Andwin Scientific B01122
Encompass_Preferred
The Biomek Span-8 P1000 Tips are compatible with Biomek NX / NXP and FX / FXP automation systems with Span-8 pods. The tips feature a clear, non-conductive version so users can easily view the aspirated sample volumes, giving you visual confirmation and added confidence during method development. This product is ideal for large volume media transfers for cellular feed and seed applications, nucleic acid extraction and purification, primer annealing for Next Generation Sequencing sample preparation, and accessing deep labware, such as 15 mL and 50 mL centrifugation tubes and deepwell plates. The tips are free of RNase, DNase, DNA, pyrogen, endotoxins, and trace metals. This is sold in a case of 480.
Catalog No. NC0692514