Learn More
Enterprise Technology Solutions Sparco Handheld Magnifier, 2X Main, 5-3/4"W x 9-3/4"H
SDP
Supplier:  Enterprise Technology Solutions SPR10601
Encompass_Preferred
High-powered magnifier is great for up-close viewing of fine print in stock listings, newspapers, maps, computer printouts, photographs, drawings and all fine prints. Magnifier is made with a full scratch-resistant, distortion-free, acrylic lens. Equipped with Fresnel lens technology, this 2X lens offers a viewing area of 5" x 8-7/8". The 3/4" diameter circular area offers 4X magnification. Inches and centimeters are imprinted on the sides of the magnifier for easy measuring.
Numéro de catalogue. 50-008-27178
Ajouter au panier
missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.