Learn More
Electron Microscopy Sciences Spare Mortar Bowl
SDP
Supplier:  Electron Microscopy Sciences 64471
Encompass_Preferred
Stainless steel motar is cooled over a bowl of liquid nitrogen. For use with Liquid Nitrogen-Cooled Mortar.
Catalog No. 50-269-62