Learn More
MSC Mitutoyo, DISPLAY UNIT SPARE PARTS
Supplier: MSC 77018570
The D-EV display unit facilitates the setup and operation of the EV counter without the need for a personal computer or additional equipment. This unit enables direct control and monitoring of measurement data, providing clear status and judgment results for industrial applications.
- Sign plus 6-digit display (with 8-digit internal operation)
- Four operation switches for channel switching, measurement mode selection, parameter setting, presetting, and tolerance configuration
- Integrated 3-color LED for channel and judgment results, 2-color status indicator, and measurement mode indicators
- RS link connectors provided for IN and OUT communication
- Operates on 12 to 24 V DC power supply
- Operating range of 0°C to 40°C
- External dimensions of 96 mm x 48 mm x 84.6 mm