Learn More
Medchemexpress LLC Sparfosic acid trisodium | 70962-66-2 | MFCD32693867 | 98.0% | 321.06 g/mol | C6H7NNa3O8P | 5 MG
SDP
Supplier:  Medchemexpress LLC HY112732B5MG
Encompass_Preferred
Sparfosic acid trisodium (CAS 70962-66-2) is a research-grade DNA antimetabolite that potently inhibits aspartate transcarbamoyl transferase (ATCase). Supplied as the trisodium salt, it is used in biochemical and cell-based studies to probe pyrimidine biosynthesis and to assess synergistic cytotoxic effects in combination with chemotherapeutic agents.
  • Potent ATCase inhibitor for studying pyrimidine biosynthesis.
  • 98.0% purity suitable for research applications.
  • Supplied as the trisodium salt in solid powder form.
  • Available in small pack sizes (5 mg-100 mg) and as 10 mM in DMSO solution.
  • Molecular weight 321.06 g/mol; formula C6H7NNa3O8P.
  • Useful in cell-based assays to evaluate synergy with chemotherapeutics.
Catalog No. 50-002-63176